Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076223.2 | NP_001069691.2 | 4 - 5 | 1176 | 80 | |
GenBank mRNA | BC116117.1 | - | 4 - 5 | 1000 | 80 | |
DQ922947.1 | - | 4 - 5 | 1176 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |