Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHYHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076243.1 | NP_001069711.1 | 9 - 10 | 773 | 107 | |
GenBank mRNA | BC122727.1 | - | 9 - 10 | 773 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Inventoried Probe spans exons |