Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076296.1 | NP_001069764.1 | 3 - 4 | 399 | 73 | |
GenBank mRNA | BC119926.1 | - | 3 - 4 | 399 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |