Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM39A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076360.2 | NP_001069828.1 | 2 - 3 | 468 | 125 | |
GenBank mRNA | BC120195.1 | - | 2 - 3 | 469 | 125 | |
BT030725.1 | - | 2 - 3 | 471 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |