Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM39A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076360.2 | NP_001069828.1 | 5 - 6 | 926 | 70 | |
GenBank mRNA | BC120195.1 | - | 5 - 6 | 927 | 70 | |
BT030725.1 | - | 5 - 6 | 929 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |