Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPIFA2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076503.1 | NP_001069971.1 | 2 - 3 | 421 | 78 | |
GenBank mRNA | DQ835286.1 | - | 2 - 3 | 421 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |