Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADCK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076895.1 | NP_001070363.1 | 6 - 7 | 814 | 93 | |
GenBank mRNA | BC123872.1 | - | 6 - 7 | 814 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |