Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076916.2 | NP_001070384.2 | 2 - 3 | 985 | 91 | |
GenBank mRNA | BC123591.1 | - | 2 - 3 | 986 | 91 | |
DN281445.1 | - | 1 - 2 | 114 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |