Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077954.1 | NP_001071422.1 | 2 - 3 | 636 | 117 | |
GenBank mRNA | BC120329.1 | - | 2 - 3 | 636 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |