Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC46A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079585.1 | NP_001073053.1 | 2 - 3 | 1102 | 70 | |
GenBank mRNA | AY999968.1 | - | 2 - 3 | 1144 | 70 | |
BC122640.1 | - | 2 - 3 | 1102 | 70 | ||
BT029834.1 | - | 2 - 3 | 1072 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |