Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMMDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080235.2 | NP_001073704.1 | 1 - 2 | 407 | 70 | |
GenBank mRNA | BC122830.1 | - | 1 - 2 | 408 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |