Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GFAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174065.2 | NP_776490.2 | 2 - 3 | 520 | 99 | |
GenBank mRNA | BC119892.1 | - | 2 - 3 | 520 | 99 | |
Y08255.1 | - | 515 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |