Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174532.3 | NP_776957.2 | 6 - 7 | 606 | 152 | |
GenBank mRNA | AF426743.1 | - | 5 - 6 | 520 | 152 | |
BC122622.1 | - | 6 - 7 | 606 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |