Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045881.2 | NP_001039346.2 | 17 - 18 | 2885 | 124 | |
GenBank mRNA | AY693782.1 | - | 8 - 9 | 850 | 124 | |
BC105482.1 | - | 17 - 18 | 2887 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |