Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045881.2 | NP_001039346.2 | 3 - 4 | 354 | 105 | |
GenBank mRNA | BC105482.1 | - | 3 - 4 | 355 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |