Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCNT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077919.2 | NP_001071387.1 | 3 - 4 | 532 | 69 | |
GenBank mRNA | AB259013.1 | - | 3 - 4 | 520 | 69 | |
BC149202.1 | - | 3 - 4 | 505 | 69 | ||
EV657636.1 | - | 3 - 4 | 522 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |