Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THG1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080233.2 | NP_001073702.1 | 3 - 4 | 556 | 118 | |
GenBank mRNA | BC122826.1 | - | 3 - 4 | 559 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |