Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: U2AF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080268.2 | NP_001073737.1 | 5 - 6 | 387 | 85 | |
GenBank mRNA | BC126638.1 | - | 5 - 6 | 388 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |