Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080354.2 | NP_001073823.1 | 5 - 6 | 374 | 98 | |
GenBank mRNA | BC126696.1 | - | 5 - 6 | 375 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |