Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WASF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081511.1 | NP_001074980.1 | 7 - 8 | 974 | 90 | |
GenBank mRNA | BC133284.1 | - | 7 - 8 | 974 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |