Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPDEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081578.1 | NP_001075047.1 | 5 - 6 | 1258 | 57 | |
GenBank mRNA | AY862876.1 | - | 3 - 4 | 284 | 57 | |
BC133366.1 | - | 5 - 6 | 1258 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |