Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XRCC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081615.2 | NP_001075084.1 | 7 - 8 | 976 | 80 | |
GenBank mRNA | BC133433.1 | - | 7 - 8 | 977 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |