Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNTNAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081745.1 | NP_001075214.1 | 11 - 12 | 1893 | 67 | |
GenBank mRNA | BT029872.1 | - | 11 - 12 | 1893 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |