Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082423.1 | NP_001075892.1 | 3 - 4 | 506 | 68 | |
GenBank mRNA | AB099087.1 | - | 3 - 4 | 438 | 68 | |
BC133497.1 | - | 3 - 4 | 506 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |