Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSNARE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082446.2 | NP_001075915.1 | 6 - 7 | 934 | 150 | |
GenBank mRNA | BC133610.1 | - | 6 - 7 | 936 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |