Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRSF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083398.1 | NP_001076867.1 | 2 - 3 | 352 | 69 | |
GenBank mRNA | BC114699.1 | - | 2 - 3 | 352 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |