Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083447.1 | NP_001076916.1 | 7 - 8 | 1224 | 108 | |
GenBank mRNA | BC133518.1 | - | 7 - 8 | 1224 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |