Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM38A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083463.1 | NP_001076932.1 | 5 - 6 | 685 | 96 | |
GenBank mRNA | BC123825.1 | - | 5 - 6 | 685 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |