Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083527.1 | NP_001076996.1 | 5 - 6 | 936 | 90 | |
GenBank mRNA | BC123648.1 | - | 5 - 6 | 936 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |