Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GFRA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083747.1 | NP_001077216.1 | 5 - 6 | 929 | 77 | |
GenBank mRNA | BC134488.1 | - | 5 - 6 | 929 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |