Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GFRA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083747.1 | NP_001077216.1 | 7 - 8 | 1147 | 60 | |
GenBank mRNA | BC134488.1 | - | 7 - 8 | 1147 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |