Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RMI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083765.1 | NP_001077234.1 | 3 - 4 | 341 | 145 | |
GenBank mRNA | BC134472.1 | - | 3 - 4 | 341 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |