Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097572.1 | NP_001091041.1 | 2 - 3 | 163 | 92 | |
GenBank mRNA | BC140658.1 | - | 2 - 3 | 163 | 92 | |
BT030694.1 | - | 2 - 3 | 201 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |