Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR1I2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103226.1 | NP_001096696.1 | 3 - 4 | 535 | 73 | |
GenBank mRNA | AY789647.1 | - | 1 - 2 | 131 | 73 | |
BC133405.1 | - | 3 - 4 | 535 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |