Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105322.1 | NP_001098792.1 | 2 - 3 | 523 | 77 | |
GenBank mRNA | BC142137.1 | - | 2 - 3 | 523 | 77 | |
BT029882.1 | - | 1 - 2 | 498 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |