Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KNG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113277.1 | NP_001106748.1 | 8 - 9 | 1196 | 73 | |
NM_175774.3 | NP_786968.2 | 8 - 9 | 1130 | 73 | ||
GenBank mRNA | BC105499.1 | - | 8 - 9 | 1131 | 73 | |
V00426.1 | - | 1130 | 73 | |||
V00427.1 | - | 1161 | 73 | |||
V01491.1 | - | 8 - 9 | 1113 | 73 | ||
V01492.1 | - | 1196 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |