Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173896.2 | NP_776321.2 | 4 - 5 | 477 | 116 | |
GenBank mRNA | AF140597.1 | - | 4 - 5 | 423 | 116 | |
BC133541.1 | - | 4 - 5 | 477 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |