Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173946.2 | NP_776371.1 | 3 - 4 | 705 | 106 | |
GenBank mRNA | AF017339.1 | - | 3 - 4 | 705 | 106 | |
BC102526.1 | - | 2 - 3 | 360 | 106 | ||
BT030567.1 | - | 2 - 3 | 349 | 106 | ||
M37974.1 | - | 3 - 4 | 692 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |