Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174077.5 | NP_776502.1 | 3 - 4 | 441 | 93 | |
NR_138142.2 | 2 - 3 | 287 | 93 | |||
GenBank mRNA | BC149266.1 | - | 3 - 4 | 420 | 93 | |
DT856720.1 | - | 3 - 4 | 418 | 93 | ||
DT861549.1 | - | 3 - 4 | 411 | 93 | ||
DV837910.1 | - | 3 - 4 | 419 | 93 | ||
DV838871.1 | - | 2 - 3 | 288 | 93 | ||
DV850873.1 | - | 2 - 3 | 266 | 93 | ||
EV673311.1 | - | 2 - 3 | 269 | 93 | ||
EV679018.1 | - | 3 - 4 | 421 | 93 | ||
L10325.1 | - | 3 - 4 | 366 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |