Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001367627.1 | NP_001354556.1 | 2 - 3 | 257 | 54 | |
NM_001367628.1 | NP_001354557.1 | 3 - 4 | 476 | 54 | ||
NM_001367629.1 | NP_001354558.1 | 4 - 5 | 693 | 54 | ||
NM_001367630.1 | NP_001354559.1 | 3 - 4 | 657 | 54 | ||
NM_001367631.1 | NP_001354560.1 | 2 - 3 | 747 | 54 | ||
NM_001367632.1 | NP_001354561.1 | 2 - 3 | 284 | 54 | ||
NM_174087.4 | NP_776512.2 | 4 - 5 | 733 | 54 | ||
GenBank mRNA | AB099033.1 | - | 1 - 2 | 70 | 54 | |
AY603685.1 | - | 3 - 4 | 599 | 54 | ||
BC116039.1 | - | 2 - 3 | 747 | 54 | ||
BC126514.1 | - | 3 - 4 | 436 | 54 | ||
DQ298746.1 | - | 2 - 3 | 259 | 54 | ||
EE251352.1 | - | 3 - 4 | 417 | 54 | ||
HQ703509.1 | - | 1 - 2 | 39 | 54 | ||
X53553.1 | - | 71 | 54 | |||
X53867.1 | - | 70 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |