Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RDH8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174167.2 | NP_776592.1 | 5 - 6 | 764 | 77 | |
GenBank mRNA | AF229846.2 | - | 5 - 6 | 760 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Bos taurus Probe spans exons |