Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAQR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046225.2 | NP_001039690.2 | 6 - 7 | 756 | 61 | |
GenBank mRNA | CO885827.1 | - | 1 - 2 | 55 | 61 | |
DV874964.1 | - | 3 - 4 | 248 | 61 | ||
DV879653.1 | - | 6 - 7 | 713 | 61 | ||
DV884229.1 | - | 6 - 7 | 707 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Detect Genomic DNA Bos taurus Probe spans exons |