Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPHX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075855.3 | NP_001069323.2 | 6 - 7 | 935 | 101 | |
NM_001314056.1 | NP_001300985.1 | 4 - 5 | 648 | 101 | ||
NR_132766.1 | 5 - 6 | 831 | 101 | |||
GenBank mRNA | BC120438.1 | - | 5 - 6 | 817 | 101 | |
DT720129.1 | - | 4 - 5 | 649 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |