Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARSL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097988.1 | NP_001091457.1 | 5 - 6 | 586 | 90 | |
GenBank mRNA | BC140583.1 | - | 5 - 6 | 586 | 90 | |
BT030685.1 | - | 5 - 6 | 692 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |