Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098044.1 | NP_001091513.1 | 5 - 6 | 732 | 72 | |
GenBank mRNA | BC140595.1 | - | 5 - 6 | 732 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |