Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098904.1 | NP_001092374.1 | 11 - 12 | 1282 | 77 | |
GenBank mRNA | BC146183.1 | - | 11 - 12 | 1282 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |