Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098912.1 | NP_001092382.1 | 6 - 7 | 571 | 98 | |
GenBank mRNA | BC142487.1 | - | 6 - 7 | 571 | 98 | |
BC151490.1 | - | 6 - 7 | 585 | 98 | ||
BC151824.1 | - | 6 - 7 | 571 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |