Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPCAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098964.2 | NP_001092434.1 | 4 - 5 | 884 | 90 | |
GenBank mRNA | BC142529.1 | - | 4 - 5 | 885 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |