Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP4A22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098990.1 | NP_001092460.1 | 10 - 11 | 1324 | 81 | |
GenBank mRNA | BC142395.1 | - | 10 - 11 | 1324 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |