Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP4A22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098990.1 | NP_001092460.1 | 12 - 13 | 1579 | 107 | |
GenBank mRNA | BC142395.1 | - | 12 - 13 | 1579 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |